Assay Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019099.4 | NP_061972.1 | 2 | 3550 | 104 | |
| NM_198926.2 | NP_945120.1 | 2 | 3408 | 104 | ||
| XM_011541783.2 | XP_011540085.1 | 4 | 3646 | 104 | ||
| GenBank mRNA | AK092074.1 | - | 1 | 945 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAP1A, member of RAS oncogene family |
RAP1A |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
-6.1kb | Anti-sense | Chr. 1 - 111542223 - 111716695 on NCBI Build GRCh38 |
| Downstream | FAM212B antisense RNA 1 | FAM212B-AS1 | 14.4kb | Anti-sense | Chr. 1 - 111739841 - 111747798 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002990.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046414.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046414.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046414.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046414.hg.1 | TC01002990.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046415.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046415.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046415.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046415.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046415.hg.1 | TC01002990.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213966.hg.1 | TC0100015284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213965.hg.1 | TC0100015284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213967.hg.1 | TC0100015284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213968.hg.1 | TC0100015284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 110188 | - | - |

FAM212B
Genomic Map