Assay Details
| Gene Symbol: INPP5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093261.1 | - | 1 | 847 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | metal regulatory transcription factor 1 | MTF1 |
MTF-1, ZRF |
-140.0 | Anti-sense | Chr. 1 - 37809567 - 37859624 on NCBI Build GRCh38 |
| Downstream | splicing factor 3a subunit 3 | SF3A3 |
PRP9, PRPF9, SAP61, SF3a60 |
95.2kb | Anti-sense | Chr. 1 - 37956975 - 37990110 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

INPP5B
Genomic Map