Assay Details
| Gene Symbol: SMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020393.1 | - | 1 | 648 | 92 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AI061139.1 | - | 1 | Chr. 9 - 70314901 - 70315307 [-] on Build GRCh38 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SMC5 antisense RNA 1 (head to head) | SMC5-AS1 | MAMDC2-AS2 | -55.5kb | Anti-sense | Chr. 9 - 70216059 - 70258874 on NCBI Build GRCh38 |
| Downstream | Kruppel like factor 9 | KLF9 |
BTEB, BTEB1 |
68.5kb | Anti-sense | Chr. 9 - 70384597 - 70414657 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SMC5
Genomic Map