Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020640.1 | - | 1 - 2 | 906 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 101 | C1orf101 | -12.8kb | Sense | Chr. 1 - 244452014 - 244640362 on NCBI Build GRCh38 | |
| Downstream |
COX20, cytochrome c oxidase assembly factor |
COX20 | FAM36A | 178.8kb | Sense | Chr. 1 - 244835306 - 244845063 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |

DESI2
Genomic Map