Assay Details
| Gene Symbol: KIAA1652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB051439.1 | - | 2 | 2555 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein subunit beta 1 like | GNB1L |
DGCRK3, FKSG1, GY2, WDR14, WDVCF |
-34.0kb | Sense | Chr. 22 - 19788411 - 19854939 on NCBI Build GRCh38 |
| Downstream | catechol-O-methyltransferase | COMT | HEL-S-98n | 46.8kb | Anti-sense | Chr. 22 - 19941740 - 19969975 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KIAA1652
Genomic Map