Assay Details
| Gene Symbol: LOC100128226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB128931.1 | - | 1 | 655 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928847 | LOC101928847 | -117.3kb | Anti-sense | Chr. 11 - 112959279 - 112963460 on NCBI Build GRCh38 | |
| Downstream | NCAM1 antisense RNA1 | NCAM1-AS1 | 187.9kb | Anti-sense | Chr. 11 - 113269532 - 113273901 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB128931.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128226
Genomic Map