Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022289.1 | - | 2 | 1753 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rap guanine nucleotide exchange factor 1 | RAPGEF1 |
C3G, GRF2 |
-141.9kb | Sense | Chr. 9 - 131576770 - 131740068 on NCBI Build GRCh38 |
| Downstream | netrin G2 | NTNG2 |
LHLL9381, Lmnt2, NTNG1, bA479K20.1 |
278.0kb | Anti-sense | Chr. 9 - 132161234 - 132244534 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MED27
Genomic Map