Assay Details
| Gene Symbol: G3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091592.1 | - | 2 | 2047 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100652758 | LOC100652758 | -16.4kb | Anti-sense | Chr. 5 - 151769793 - 151772176 on NCBI Build GRCh38 | |
| Downstream | glycine receptor alpha 1 | GLRA1 |
HKPX1, STHE |
32.4kb | Anti-sense | Chr. 5 - 151820798 - 151924836 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000356245.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000394123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000507878.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000517947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000518726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000518726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000518726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000519832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520177.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000520578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000522761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000523519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | ENST00000627077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_005754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009131.hg.1 | NM_198395.1 | Refseq |

G3BP1
Genomic Map