Assay Details
| Gene Symbol: TTLL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123328.1 | - | 2 | 1251 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dual specificity phosphatase 15 | DUSP15 |
C20orf57, VHY |
-71.0kb | Anti-sense | Chr. 20 - 31845578 - 31870744 on NCBI Build GRCh38 |
| Downstream | p53 and DNA damage regulated 1 | PDRG1 |
C20orf126, PDRG |
1.6kb | Anti-sense | Chr. 20 - 31944955 - 31952080 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TTLL9
Genomic Map