Assay Details
| Gene Symbol: LOC100128697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124043.1 | - | 2 | 1851 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR202 host gene | MIR202HG | -16.2kb | Sense | Chr. 10 - 133246479 - 133247885 on NCBI Build GRCh38 | |
| Downstream | tubulin gamma complex associated protein 2 | TUBGCP2 |
GCP-2, GCP2, Grip103, SPBC97, Spc97p, h103p, hGCP2, hSpc97 |
13.8kb | Sense | Chr. 10 - 133278630 - 133312337 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128697
Genomic Map