Assay Details
| Gene Symbol: TRIM36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125426.1 | - | 1 | 2152 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927078 | LOC101927078 | -404.1kb | Sense | Chr. 5 - 114447418 - 114773413 on NCBI Build GRCh38 | |
| Downstream | protein geranylgeranyltransferase type I subunit beta | PGGT1B |
BGGI, GGTI |
19.9kb | Sense | Chr. 5 - 115200247 - 115262887 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TRIM36
Genomic Map