Assay Details
| Gene Symbol: SPATA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126086.1 | - | 2 | 2348 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPATA3 antisense RNA 1 (head to head) | SPATA3-AS1 | -311.0 | Anti-sense | Chr. 2 - 230984368 - 230996032 on NCBI Build GRCh38 | |
| Downstream | chromosome 2 open reading frame 72 | C2orf72 | 37.4kb | Sense | Chr. 2 - 231037566 - 231049714 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |

SPATA3
Genomic Map