| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24720938 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04533614 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
546764 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3374221 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P115973 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P115973 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
61725 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P115973 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P115973 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
168229 |
AF006464.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
168229 |
BC109098.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
168229 |
BC109099.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
168229 |
NM_005592.1 |
refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
62028 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P115973 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01417027 |
- |
- |