Assay Details
| Gene Symbol: CSAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128796.1 | - | 1 | 1453 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterol O-acyltransferase 2 | SOAT2 |
ACACT2, ACAT2, ARGP2 |
-50.0kb | Anti-sense | Chr. 12 - 53102437 - 53124539 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 740 | ZNF740 | Zfp740 | 4.3kb | Anti-sense | Chr. 12 - 53180715 - 53190870 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CSAD
Genomic Map