Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 35B |
SNORD35B |
RNU35B, U35B |
-1.9kb | Sense | Chr. 19 - 49497719 - 49497806 on NCBI Build GRCh38 |
| Downstream | Fc fragment of IgG receptor and transporter | FCGRT |
FCRN, alpha-chain |
9.9kb | Sense | Chr. 19 - 49512279 - 49526428 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map