Assay Details
| Gene Symbol: LOC340340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040593.1 | - | 3 | 2645 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 71-like | CCDC71L | C7orf74 | -118.8kb | Anti-sense | Chr. 7 - 106656765 - 106661188 on NCBI Build GRCh38 |
| Downstream | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma | PIK3CG |
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K |
82.4kb | Sense | Chr. 7 - 106865278 - 106908978 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671171 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16002746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16002755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927864 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671130 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05671129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25927876 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 750925 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 399609 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3531373 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2915 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 114320 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16210901 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38144787 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38144787 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P16210901 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1561254_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561254_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561254_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561254_at11 | - | - |
| Human Genome U95 Set | 50406_PM_at | - | - |
| Human Genome U95 Set | 50406_at | - | - |

LOC340340
Genomic Map