Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methyltransferase like 7B | METTL7B | ALDI | -3.8kb | Sense | Chr. 12 - 55681546 - 55684611 on NCBI Build GRCh38 |
| Downstream | biogenesis of lysosomal organelles complex 1 subunit 1 | BLOC1S1 |
BLOS1, BORCS1, GCN5L1, MICoA, RT14 |
24.9kb | Sense | Chr. 12 - 55716034 - 55719707 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map