Assay Details
| Gene Symbol: G3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X78262.1 | - | 1 | 3648 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100652758 | LOC100652758 | -601.0 | Anti-sense | Chr. 5 - 151769793 - 151772176 on NCBI Build GRCh38 | |
| Downstream | glycine receptor alpha 1 | GLRA1 |
HKPX1, STHE |
43.5kb | Anti-sense | Chr. 5 - 151820798 - 151924836 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

G3BP1
Genomic Map