Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MOB kinase activator 3A | MOB3A |
MOB1C, MOBKL2A, moblak |
Anti-sense | Chr. 19 - 2071036 - 2096502 on NCBI Build GRCh38 | |
| Downstream | IZUMO family member 4 | IZUMO4 |
C19orf36, IMAGE:4215339 |
7.3kb | Sense | Chr. 19 - 2096848 - 2099590 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map