Assay Details
| Gene Symbol: TCEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094425.1 | - | 1245 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of G-protein signaling 20 | RGS20 |
RGSZ1, ZGAP1, g(z)GAP, gz-GAP |
-31.3kb | Sense | Chr. 8 - 53851808 - 53959304 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105375839 | LOC105375839 | 52.0kb | Anti-sense | Chr. 8 - 54042987 - 54045846 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TCEA1
Genomic Map