Assay Details
| Gene Symbol: MGC10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC004943.1 | - | 1 | 348 | 3613095 | 87 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ceramide synthase 1 | CERS1 |
EPM8, LAG1, LASS1, UOG1 |
Sense | Chr. 19 - 18868546 - 18896727 on NCBI Build GRCh38 | |
| Downstream | coatomer protein complex subunit epsilon | COPE | epsilon-COP | 17.4kb | Sense | Chr. 19 - 18899514 - 18919464 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC004943.1 transcripts.
Set Membership: |
|

MGC10814
Genomic Map