Assay Details
| Gene Symbol: EMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017854.1 | - | 1 | 150 | 101 | |
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK309521.1 | - | 1371 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | germ cell associated 1 | GSG1 | -93.1kb | Anti-sense | Chr. 12 - 13083537 - 13103720 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1559 | LINC01559 | C12orf36 | 173.6kb | Anti-sense | Chr. 12 - 13370671 - 13376745 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EMP1
Genomic Map