Assay Details
| Gene Symbol: VPS13A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026668.2 | 1 - 2 | 298 | 82 | ||
| GenBank mRNA | AA626167.1 | - | 1 - 2 | 82 | ||
| DB509755.1 | - | 1 - 2 | 304 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box B2 | FOXB2 | bA159H20.4 | -155.8kb | Anti-sense | Chr. 9 - 77019655 - 77020953 on NCBI Build GRCh38 |
| Downstream | G protein subunit alpha 14 | GNA14 | 245.2kb | Sense | Chr. 9 - 77423079 - 77648316 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001225.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3318243 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4704 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4703 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3318243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16228761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27091796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06753108 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753109 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753106 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16228748 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753090 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753134 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753104 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16228751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753101 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753095 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753136 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753113 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753103 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16228752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753128 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753116 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753139 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753111 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753130 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06753092 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 240767_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015749.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015749.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015749.hg.1 | TC09001225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010480.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010480.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015748.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015748.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015748.hg.1 | TC09001225.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010480.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010480.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010480.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171699.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171699.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171699.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171699.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171699.hg.1 | TC0900010480.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900171698.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171698.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171698.hg.1 | ENST00000415172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171698.hg.1 | NR_026668.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171698.hg.1 | TC0900010480.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037598 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037598 | - | - |

VPS13A-AS1
Genomic Map