Assay Details
| Gene Symbol: RPS2P32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026676.1 | 1 | 855 | 92 | ||
| GenBank mRNA | BC026177.1 | - | 1 | 664 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | insulin like growth factor 2 mRNA binding protein 3 | IGF2BP3 |
CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3 |
-19.9kb | Anti-sense | Chr. 7 - 23310209 - 23470674 on NCBI Build GRCh38 |
| Downstream | transformer 2 alpha homolog | TRA2A |
AWMS1, HSU53209 |
13.4kb | Anti-sense | Chr. 7 - 23504780 - 23532041 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000148.hg.1
Set Membership: |
|

RPS2P32
Genomic Map