Assay Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026737.1 | 1 - 2 | 107 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PAK1 interacting protein 1 | PAK1IP1 |
MAK11, PIP1, WDR84, bA421M1.5, hPIP1 |
Anti-sense | Chr. 6 - 10690865 - 10709782 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 14C | TMEM14C |
C6orf53, HSPC194, MSTP073, NET26, bA421M1.6 |
28.3kb | Anti-sense | Chr. 6 - 10722915 - 10731129 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001265.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | ENST00000379586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | ENST00000460742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015163.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015164.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015164.hg.1 | ENST00000379586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015164.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015164.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015164.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173415.hg.1 | TC0600010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173416.hg.1 | TC0600010790.hg.1 | Clariom D TC |

C6orf52
Genomic Map