Assay Details
| Gene Symbol: MGC2889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026877.1 | 1 | 1442 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Mab-21 domain containing 2 | MB21D2 | C3orf59 | -323.6kb | Anti-sense | Chr. 3 - 192796815 - 192918161 on NCBI Build GRCh38 |
| Downstream | ATPase 13A5 | ATP13A5 | 30.8kb | Anti-sense | Chr. 3 - 193274789 - 193378824 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001064.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P519672 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P519672 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 28162 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17496 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P519672 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P519672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23006520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02866259 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P519672 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P519672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866259 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866254 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15523607 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866264 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866261 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02866257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 211718_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018621.hg.1 | NR_026877.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018621.hg.1 | TC03001064.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300171902.hg.1 | NR_026877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171902.hg.1 | NR_026877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171902.hg.1 | TC0300009891.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171901.hg.1 | NR_026877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171901.hg.1 | NR_026877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171901.hg.1 | TC0300009891.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 211718_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211718_at | - | - |
| Human Genome U133A 2.0 Array | 211718_at | - | - |
| Human Genome U133A 2.0 Array | 211718_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw593402 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211718_at2 | - | - |
| Human Genome U133 Set | 211718_at | - | - |
| Human Genome U133 Set | 211718_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023721 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023721 | - | - |

MGC2889
Genomic Map