Assay Details
| Gene Symbol: LOC100128239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027276.1 | 1 - 2 | 582 | 77 | ||
| GenBank mRNA | AK125302.1 | - | 1 - 2 | 582 | 77 | |
| AK308670.1 | - | 1 - 2 | 202 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | immunoglobulin superfamily member 9B | IGSF9B | -75.3kb | Anti-sense | Chr. 11 - 133908564 - 133956960 on NCBI Build GRCh38 | |
| Downstream | junctional adhesion molecule 3 | JAM3 |
JAM-2, JAM-3, JAM-C, JAMC |
31.3kb | Sense | Chr. 11 - 134068925 - 134151757 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001211.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11015163.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015163.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015163.hg.1 | TC11001211.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015162.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015162.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015162.hg.1 | TC11001211.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203049.hg.1 | TC1100013116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203052.hg.1 | TC1100013116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203050.hg.1 | TC1100013116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203046.hg.1 | TC1100013116.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203047.hg.1 | TC1100013116.hg.1 | Clariom D TC |

LOC100128239
Genomic Map