Assay Details
| Gene Symbol: FKBP9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027339.1 | 1 - 2 | 478 | 86 | ||
| GenBank mRNA | BM014562.1 | - | 1 - 2 | 478 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
vesicular, overexpressed in cancer, prosurvival protein 1 |
VOPP1 |
ECOP, GASP |
-115.3kb | Sense | Chr. 7 - 55434406 - 55572520 on NCBI Build GRCh38 |
| Downstream | putative uncharacterized protein FLJ44672 | LOC101060341 | 57.0kb | Sense | Chr. 7 - 55746453 - 55750715 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001398.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021781.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021781.hg.1 | TC07001398.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021780.hg.1 | ENST00000455909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021780.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021780.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07021780.hg.1 | TC07001398.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179680.hg.1 | TC0700011151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000441699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | ENST00000455909.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_003949.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027340.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011151.hg.1 | NR_027342.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179682.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179682.hg.1 | NR_027339.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700179682.hg.1 | TC0700011151.hg.1 | Clariom D TC |

FKBP9P1
Genomic Map