Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027398.1 | 1 - 2 | 209 | 83 | ||
| GenBank mRNA | BC000757.2 | - | 1 - 2 | 162 | 2820364 | 83 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 16 open reading frame 86 | C16orf86 | -5.8kb | Anti-sense | Chr. 16 - 67666806 - 67670523 on NCBI Build GRCh38 | |
| Downstream | RAN binding protein 10 | RANBP10 | 3.7kb | Sense | Chr. 16 - 67723066 - 67806652 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001195.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC000757.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016616.hg.1 | ENST00000268797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016616.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016616.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016616.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016616.hg.1 | TC16001195.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016614.hg.1 | ENST00000268797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016614.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016614.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016614.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016614.hg.1 | TC16001195.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | TC1600010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182149.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | TC1600010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182128.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010649.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NR_027398.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | TC1600010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000268797.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602279.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | ENST00000602855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NM_001243650.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NM_030819.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600182143.hg.1 | NR_027398.1 | Refseq |

GFOD2
Genomic Map