Assay Details
| Gene Symbol: RNF216P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023385.1 | 4 - 5 | 581 | 65 | ||
| GenBank mRNA | AK303831.1 | - | 4 - 5 | 432 | 65 | |
| AK308526.1 | - | 4 - 5 | 516 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | monocyte to macrophage differentiation associated 2 | MMD2 | PAQR10 | -25.7kb | Anti-sense | Chr. 7 - 4892245 - 4959213 on NCBI Build GRCh38 |
| Downstream | RB associated KRAB zinc finger | RBAK | ZNF769 | 47.7kb | Sense | Chr. 7 - 5045821 - 5069488 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

RNF216P1
Genomic Map