| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019123.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019123.hg.1 |
TC10001521.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019122.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019122.hg.1 |
TC10001521.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019121.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019121.hg.1 |
TC10001521.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000177468.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177468.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177468.hg.1 |
TC1000011408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177472.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177472.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177472.hg.1 |
TC1000011408.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177469.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177469.hg.1 |
NR_024467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177469.hg.1 |
TC1000011408.hg.1 |
Clariom D TC |