Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371397 | LOC105371397 | Sense | Chr. 16 - 87700832 - 87703017 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC102724467 | LOC102724467 | 64.6kb | Sense | Chr. 16 - 87766331 - 87779148 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map