Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1395 | LINC01395 | -80.4kb | Sense | Chr. 11 - 129612116 - 129617269 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 45B | TMEM45B | 117.5kb | Anti-sense | Chr. 11 - 129815781 - 129860003 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map