Assay Details
| Gene Symbol: DCAF8L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136533.1 | NP_001130005.1 | 1 | 347 | 101 | |
| GenBank mRNA | BC157859.1 | - | 1 | 347 | 9057078 | 101 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000131.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 394103 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3310009 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1893 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35936 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3310009 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3310009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364911 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364935 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364912 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364912 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364911 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364935 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11659493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17328506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32364929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659518 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17328501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659475 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659473 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17328506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659529 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17328511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17328508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659514 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659500 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11659513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11738229_at | - | - |
| Affymetrix Human Genome U219 Array | 11738229_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001677.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001677.hg.1 | TC0X000131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001675.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X001675.hg.1 | TC0X000131.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146858.hg.1 | TC0X00006856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146859.hg.1 | TC0X00006856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00146860.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146860.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146860.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146860.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146860.hg.1 | TC0X00006856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00146855.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146855.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146855.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146855.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146855.hg.1 | TC0X00006856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000431122.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000451261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000545306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | ENST00000583068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | NM_001136533.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00146856.hg.1 | TC0X00006856.hg.1 | Clariom D TC |
| Human Genome U95 Set | 59155_at | - | - |
| Human Genome U95 Set | 59155_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040788 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 202840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812355 | BC157859.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812355 | NM_001136533.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812355 | BC157859.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812355 | NM_001136533.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812355 | BC157859.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812355 | NM_001136533.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812355 | BC157859.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812355 | NM_001136533.1 | human_transcriptome |

DCAF8L2
Genomic Map