Assay Details
| Gene Symbol: NKAIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152864.3 | NP_690603.3 | 1 - 2 | 148 | 68 | |
| XM_005260192.1 | XP_005260249.1 | 1 - 2 | 110 | 68 | ||
| XM_011528527.1 | XP_011526829.1 | 1 - 2 | 110 | 68 | ||
| GenBank mRNA | AK126246.1 | - | 1 - 2 | 71 | 68 | |
| AK225765.1 | - | 1 - 2 | 102 | 68 | ||
| BC041812.1 | - | 1 - 2 | 146 | 5302801 | 68 | |
| BG708360.1 | - | 1 - 2 | 152 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001027.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20014048.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014048.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014048.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014045.hg.1 | ENST00000370307.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014045.hg.1 | ENST00000370313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014045.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014045.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014045.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168354.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168357.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168357.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168357.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168357.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168357.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168356.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168356.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168356.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168356.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168356.hg.1 | NM_152864.3 | Refseq |

NKAIN4
Genomic Map