Assay Details
| Gene Symbol: NKAIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152864.3 | NP_690603.3 | 6 - 7 | 706 | 76 | |
| XM_017027638.1 | XP_016883127.1 | 6 - 7 | 1126 | 76 | ||
| XM_017027639.1 | XP_016883128.1 | 6 - 7 | 1112 | 76 | ||
| GenBank mRNA | AK126246.1 | - | 6 - 7 | 629 | 76 | |
| AK225765.1 | - | 6 - 7 | 660 | 76 | ||
| BC041812.1 | - | 6 - 7 | 704 | 5302801 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001027.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20014026.hg.1 | ENST00000370313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014026.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014026.hg.1 | ENST00000466885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014026.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014026.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014027.hg.1 | ENST00000370313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014027.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014027.hg.1 | ENST00000466885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014027.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014027.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | ENST00000370307.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | ENST00000370313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | ENST00000466885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014028.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014025.hg.1 | ENST00000370313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014025.hg.1 | ENST00000370316.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014025.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20014025.hg.1 | TC20001027.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168338.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370307.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000370317.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000461738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000472670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | ENST00000486495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009802.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168336.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000466885.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | ENST00000470246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168337.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | NM_152864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | TC2000009802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | ENST00000370313.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | ENST00000370316.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000168335.hg.1 | NM_152864.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 211587 | - | - |

NKAIN4
Genomic Map