Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 173 member B | FAM173B | JS-2 | -2.6kb | Anti-sense | Chr. 5 - 10225507 - 10249909 on NCBI Build GRCh38 |
| Downstream | carboxymethylenebutenolidase homolog | CMBL | JS-1 | 24.7kb | Anti-sense | Chr. 5 - 10277595 - 10308056 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map