Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1216 | LINC01216 | EMCN-IT3 | -485.4kb | Anti-sense | Chr. 4 - 100660279 - 100675113 on NCBI Build GRCh38 |
| Downstream | microRNA 8066 | MIR8066 | hsa-mir-8066 | 78.5kb | Anti-sense | Chr. 4 - 101240795 - 101240872 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map