Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GH3 domain containing | GHDC |
D11LGP1, LGP1 |
-39.3kb | Anti-sense | Chr. 17 - 42189087 - 42194532 on NCBI Build GRCh38 |
| Downstream | signal transducer and activator of transcription 5A | STAT5A |
MGF, STAT5 |
52.1kb | Sense | Chr. 17 - 42287547 - 42311943 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map