Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras homolog family member D | RHOD |
ARHD, RHOHP1, RHOM, Rho |
-123.9kb | Anti-sense | Chr. 11 - 67056818 - 67072017 on NCBI Build GRCh38 |
| Downstream | G protein-coupled receptor kinase 2 | GRK2 |
ADRBK1, BARK1, BETA-ARK1 |
69.2kb | Anti-sense | Chr. 11 - 67266420 - 67286558 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map