Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
SMG6, nonsense mediated mRNA decay factor |
SMG6 |
C17orf31, EST1A, SMG-6, hSMG5/7a |
Sense | Chr. 17 - 2059839 - 2303836 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927839 | LOC101927839 | 110.2kb | Anti-sense | Chr. 17 - 2232678 - 2233610 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map