Assay Details
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012558.1 | XP_016868047.1 | 10 | 1244 | 86 | |
| XM_017012559.1 | XP_016868048.1 | 10 | 1244 | 86 | ||
| GenBank mRNA | AJ276310.1 | - | 1 | 63 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADH:ubiquinone oxidoreductase subunit B2 | NDUFB2 |
AGGG, CI-AGGG |
-79.0kb | Anti-sense | Chr. 7 - 140696681 - 140706646 on NCBI Build GRCh38 |
| Downstream | mitochondrial ribosomal protein S33 | MRPS33 |
CGI-139, MRP-S33, PTD003, S33mt |
220.4kb | Sense | Chr. 7 - 141006161 - 141014981 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BRAF
Genomic Map