Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | taxilin beta | TXLNB |
C6orf198, LST001, MDP77, dJ522B19.2 |
Anti-sense | Chr. 6 - 139159160 - 139324011 on NCBI Build GRCh38 | |
| Downstream | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2 | CITED2 |
ASD8, MRG-1, MRG1, P35SRJ, VSD2 |
89.0kb | Anti-sense | Chr. 6 - 139372255 - 139374650 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map