Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927501 | LOC101927501 | -483.3kb | Sense | Chr. X - 43176994 - 43226598 on NCBI Build GRCh38 | |
| Downstream | monoamine oxidase B | MAOB | 55.4kb | Anti-sense | Chr. X - 43766610 - 43882475 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map