Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1237 | LINC01237 | Sense | Chr. 2 - 241881363 - 242078722 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC102723927 | LOC102723927 | 54.4kb | Sense | Chr. 2 - 241970683 - 241977276 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map