Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WW domain containing oxidoreductase | WWOX |
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1 |
Sense | Chr. 16 - 78099413 - 79212667 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371352 | LOC105371352 | 310.3kb | Anti-sense | Chr. 16 - 79202624 - 79211600 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map