Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FKBP1A-SDCBP2 readthrough (NMD candidate) | FKBP1A-SDCBP2 | Sense | Chr. 20 - 1309909 - 1393172 on NCBI Build GRCh38 | ||
| Downstream | FK506 binding protein 1A | FKBP1A |
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE |
32.1kb | Sense | Chr. 20 - 1368977 - 1393172 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map