Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1370 | LINC01370 | HI-LNC25 | -391.1kb | Anti-sense | Chr. 20 - 40004461 - 40008529 on NCBI Build GRCh38 |
| Downstream | MAF bZIP transcription factor B | MAFB |
KRML, MCTO |
284.3kb | Sense | Chr. 20 - 40685848 - 40689240 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map