Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NAALADL2 antisense RNA 2 | NAALADL2-AS2 | -76.5kb | Anti-sense | Chr. 3 - 175235092 - 175271096 on NCBI Build GRCh38 | |
| Downstream | microRNA 4789 | MIR4789 | 20.3kb | Sense | Chr. 3 - 175369540 - 175369621 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map