Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuronal growth regulator 1 | NEGR1 |
DMML2433, IGLON4, KILON, Ntra |
-63.4kb | Anti-sense | Chr. 1 - 71402942 - 72282850 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC339468 | LOC339468 | 417.0kb | Sense | Chr. 1 - 72765005 - 72783708 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map